Learn More
Bellco Glass, Inc. 1mL Pipet, Individually
SDP
Supplier:  Bellco Glass, Inc. 229201A00
Encompass_Preferred
Peel-to-open plastic/plastic wrappers are the ideal option for eliminating particulate; proven benefit over traditional paper/plastic wrapperWrappers reduce static cling versus other brands of plastic wrap pipetsAll plastic wrapper can be wiped with alcohol without smearing ink or penetrating paperAll serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934Bi-directional (positive and negative) graduations are printed in bold easy-to-read inkPremium material barrier plugColor coded stripes and packaging enable quick volume identification Pack Description100 Individually Wrapped/BagCase Quantity800SterileYesNon-pyrogenicYesPrimary ColorClearSecondary ColorYellowPrimary MaterialPolystyreneMaximum Volume1mLGraduatedYesGraduation Increments (Subdivisions)1/100mLRNase/DNase FreeYes
Catalog No. 50-232-9637