Learn More
AGRI PRO ENTERPRISES OF IOWA INC 1 ML with spike & 42" tubing
Supplier:  AGRI PRO ENTERPRISES OF IOWA INC 268018
Encompass_Preferred
1ML RING-GRIP SYRINGE W/SPIKE KITThe Ring-Grip Syringe from Prodigy Instruments is lightweight, easy to hang and the smooth injection action makes vaccinating a breeze! Features a poly barrel and blue o-ring for long acting oil based and suspension vaccines. Set required dose by aligning front of o-ring with dose graduation marking on the cylinder. Metal luerlock cap with metal nib for durability. Convenient and attractive retail packaging.
Catalog No. NC2824318