Learn More
Chemglass Life Sciences Serological Pipette, 1mL, Color Coded Yellow, Bulk Packed, Sterile (20 bags of 50)
SDP
Supplier:  Chemglass Life Sciences CGN3482001
Encompass_Preferred
Bulk packed serological pipettes are made of super clear clarity polystyrene (PS). Serological pipettes are designed to be used in both tissue culture and bacterial culture experiments, and other liquid handling applications.Edged color-coded band for identificationInclude negative graduations for extra volumeNon-pyrogenic, non-cytotoxic, non-hemolytic, DNase- and RNase-freePolyolefin fiber filter to reduce contaminationMade from high clarity polystyreneSterilized by E-beam
Catalog No. 50-189-7703