Learn More
CELLTREAT Scientific Products 1mL Serological Pipet, Individual Paper/Plastic Wrapper Packed in Cartons, Sterile pipette
SDP
Supplier:  CELLTREAT Scientific Products 229201
Encompass_Preferred
CELLTREAT Scientific Products 1mL Serological Pipet. Convenient shelf carton allows easy access to pipets and neat storage. All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934. Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink. Premium material barrier plug. Color coded stripes and packaging enable quick volume identification. Printed indicator arrow for quickly locating negative graduations when using a pipet controller. RNase/DNase Free, Non-pyrogenic. Every case is is printed with the lot number for quality traceability. Packaged sterile, 200 individually wrapped in paper/plastic wrapper per dispensor carton, 800/case.
Catalog No. 50-202-114