Learn More
Chemglass Life Sciences Tube, Vacuum Hydrolysis, 1mL Capacity, 8mm Tube OD x 60mm Tube Length
SDP
Supplier:  Chemglass Life Sciences CG402501
Encompass_Preferred
Tube, Vacuum Hydrolysis, 1mL Capacity, 8mm Tube OD x 60mm Tube Length. Tube is useful in protein hydrolysis, amino acid separations, freeze drying or any application under vacuum.
Catalog No. CG402501