Learn More
Cayman Chemical 1MyrIstoyl2hydroxysnglyc 50mg
SDP
Supplier:  Cayman Chemical 2095850 MG
Encompass_Preferred
A saturated 14:0 lysophosphatidylcholine that consists of one chain of myristic acid at the sn-1 position
Catalog No. 50-298-3244