Learn More
Cayman Chemical 1Ohexadecyl2Docosahexaeno 10mg
SDP
Supplier:  Cayman Chemical 6090310 MG
Encompass_Preferred
1-O-hexadecyl-2-Docosahexaenoyl-sn-glycero-3-PC is a PAF analog which contains docosahexaenoate at the sn-2 position rather than the acetate moiety found in PAF C-16. 1-O-hexadecyl-2-Docosahexaenoyl-sn-glycero-3-PC can be oxidatively fragmented resulting in the production of phospholipids with PAF-like activity.{1938,2654}
Catalog No. 50-289-5498