Learn More
Cayman Chemical 1Ohexadecyl2EIcosapentaen 10mg
SDP
Supplier:  Cayman Chemical 6090710 MG
Encompass_Preferred
Can be formed by incorporation of EPA into lyso-PAF C-16, as has been demonstrated using neutrophils from monkeys and humans fed a diet enriched in fish oils; serves as a substrate for PAF C-16 formation by the remodeling pathway.
Catalog No. 50-289-6125