Learn More
Cayman Chemical 1Oleoyl2PalmItoylracglyc 10mg
SDP
Supplier:  Cayman Chemical 2716610 MG
Encompass_Preferred
A diacylglycerol; enhances the inhibition of superoxide anion production by cytochalasin B at 10 µM
Catalog No. 50-293-7872