Learn More
Cayman Chemical 1Oleoyl2PalmItoylsnglyce 10mg
SDP
Supplier:  Cayman Chemical 2601210 MG
Encompass_Preferred
A phospholipid; found in biological membranes; can be used in the generation of liposomes and other artificial membranes
Catalog No. 50-293-5765