Learn More
Cayman Chemical 1Oxyl2 2 5 5tetramthylpyr 10mg
SDP
Supplier:  Cayman Chemical 1614810 MG
Encompass_Preferred
An N-hydroxysuccinimide ester of the nitroxide spin label tempyo; an active acylating agent that preferentially targets α-amino groups, used to spin label proteins and aminoacylated RNA species
Catalog No. 50-297-0433