Learn More
Cayman Chemical 1PalmItol2Docosahexaenol 10mg
SDP
Supplier:  Cayman Chemical 3344010 MG
Encompass_Preferred
A phospholipid; found in the nucleus, but not the mitochondria, of mouse liver; levels are lower in pre-term infant plasma compared with maternal and fetal plasma
Catalog No. 50-294-7732