Learn More
Cayman Chemical 1PalmItoyl2Oleoyl3LInole 10mg
SDP
Supplier:  Cayman Chemical 6266010 MG
Encompass_Preferred
POL is a common triglyceride component in seed and vegetable oils including olive, sesame, soybean, canola, corn, hazelnut, and many others. POL is one of the standard triglyceride components used for the USP analysis of sesame oil for pharmaceutical applications.{8754}
Catalog No. 50-289-5967