Learn More
Cayman Chemical 1PalmItoyl2Oleoylsnglyce 10mg
SDP
Supplier:  Cayman Chemical 2006810 MG
Encompass_Preferred
A phospholipid; has been used in the synthesis of lipid membranes and synthetic high-density lipoproteins
Catalog No. 50-298-2555