Learn More
Cayman Chemical 1PalmItoyl2Oleoylsnglyce 5mg
SDP
Supplier:  Cayman Chemical 209635 MG
Encompass_Preferred
A phospholipid containing saturated palmitic acid (16:0) and monounsaturated oleic acid (18:1) inserted at the sn-1 and sn-2 positions, respectively
Catalog No. 50-298-1744