Learn More
Cayman Chemical 1PalmItoyl2Stearoyl3Oleo 5mg
SDP
Supplier:  Cayman Chemical 270685 MG
Encompass_Preferred
A triacylglycerol; has been detected in RAW 264.7 cells by neutral loss MS; increased serum levels are a potential biomarker for NAFLD
Catalog No. 50-293-9730