Learn More
Cayman Chemical 1PalmItoyld92ArachIdonyl 100ug
SDP
Supplier:  Cayman Chemical 29342100 UG
Encompass_Preferred
An internal standard for the quantification of 1,3-dipalmitoyl-2-arachidonoyl-sn-glycerol by GC- or LC-MS
Catalog No. 50-299-0579