Learn More
Cayman Chemical 1Pentadecnoylracglycerol 50mg
SDP
Supplier:  Cayman Chemical 2695950 MG
Encompass_Preferred
A monoacylglycerol; has been found in wheat bran extracts; levels are increased in a HepaRG cell-based model of hepatic steatosis induced by BSA-conjugated palmitate
Catalog No. 50-293-9728