Learn More
Strem, An Ascensus Company CAS# 138517-61-0. 2g. (1R,2R)-(+)-1,2-Diaminocyclohexane-N,N'-bis(2'-diphenylphosphinobenzoyl), 98% (R,R)-DACH-Phenyl Trost Ligand. MFCD01631273
SDP
Supplier:  Strem, An Ascensus Company 1509602G
Encompass
CAS# 138517-61-0. 2g. (1R,2R)-(+)-1,2-Diaminocyclohexane-N,N'-bis(2'-diphenylphosphinobenzoyl), 98% (R,R)-DACH-Phenyl Trost Ligand. MFCD01631273. Molecular Weight: 690.76. Molecular Formula: C44H40N2O2P2. Color/form: white to off-white pwdr. Strem# 15-0960. http://www.strem.com/catalog/v/15-0960/
Catalog No. 50-901-09988