Learn More
Strem, An Ascensus Company CAS# 174810-09-4. 2g. (1R,2R)-(+)-1,2-Diaminocyclohexane-N,N'-bis(2-diphenylphosphino-1-naphthoyl), min. 94% (R,R)-DACH-Naphthyl Trost Ligand. MFCD02684551
SDP
Supplier:  Strem, An Ascensus Company 1509632G
Encompass
CAS# 174810-09-4. 2g. (1R,2R)-(+)-1,2-Diaminocyclohexane-N,N'-bis(2-diphenylphosphino-1-naphthoyl), min. 94% (R,R)-DACH-Naphthyl Trost Ligand. MFCD02684551. Molecular Weight: 790.88. Molecular Formula: C52H44N2O2P2. Color/form: off-white pwdr. Strem# 15-0963. http://www.strem.com/catalog/v/15-0963/
Catalog No. 50-901-09994