Learn More
Strem, An Ascensus Company CAS# 35132-20-8. 500mg. (1R,2R)-(+)-1,2-Diphenylethylenediamine, min. 97% (R,R)-DPEN. MFCD00082769
SDP
Supplier:  Strem, An Ascensus Company 070474500MG
Encompass
CAS# 35132-20-8. 500mg. (1R,2R)-(+)-1,2-Diphenylethylenediamine, min. 97% (R,R)-DPEN. MFCD00082769. Molecular Weight: 212.29. Molecular Formula: (C6H5)(NH2)CHCH(NH2)(C6H5). Color/form: white to pale yellow xtl. Strem# 07-0474. http://www.strem.com/catalog/v/07-0474/
Catalog No. 50-901-09005