Learn More
Medchemexpress LLC (1R,2R)-2-(pyridin-3-yl)cyclopropane-1-carboxylic acid | 1423769-08-7 | MFCD22419060 | 163.18 g/mol | C9H9NO2 | 5 G

Supplier: Medchemexpress LLC HYW0639715G
(1R,2R)-2-(pyridin-3-yl)cyclopropane-1-carboxylic acid is a chiral cyclopropane carboxylic acid intermediate used in medicinal chemistry research. Supplied as a powder for research use.
- Chiral cyclopropane carboxylic acid intermediate.
- CAS number 1423769-08-7.
- Molecular formula C9H9NO2.
- Molecular weight 163.18 g/mol.
- Supplied as powder suitable for synthetic applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.