Learn More
Medchemexpress LLC HY-134819A 5mg , (1R,2R)-ML-SI3 CAS:2418594-00-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY134819A5MG
Encompass_Preferred
HY-134819A 5mg (1R,2R)-ML-SI3 CAS: 2418594-00-8 (1R,2R)-ML-SI3 is a potent inhibitor of both TRPML1 and TRPML2 (IC50 values of 1.6 and 2.3 μM) and a weak inhibitor (IC50 12.5 μM) of TRPML3. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0066