Learn More
Medchemexpress LLC (1R,2R)-N,N'-bis(phenylmethyl)-1,2-cyclohexanediamine | 143443-23-6 | 97.7% | 294.44 g/mol | C20H26N2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW037146100MG
Encompass_Preferred
(1R,2R)-N1,N2-Dibenzylcyclohexane-1,2-diamine is a chiral diamine used as a drug intermediate in the synthesis of active pharmaceutical compounds and research chemicals. It is supplied in small-scale quantities for laboratory research and is accompanied by product and safety data sheets.
  • Chiral diamine useful for asymmetric synthesis and ligand development.
  • Acts as an intermediate in the preparation of APIs and bioactive molecules.
  • Offered in multiple small-scale pack sizes for research and development.
  • Characterized by molecular formula C20H26N2 and molecular weight 294.44 g/mol.
  • Provided with a product data sheet and an SDS for handling and safety information.
Catalog No. 50-002-99796