Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (1R,2S)-2-[(3,5-dichlorophenyl)carbamoyl]cyclohexane-1-carboxylic acid | 1263273-14-8 | MFCD26142602 | 95.1% | C14H15Cl2NO3 | 10MG
SDP

Supplier:  Medchemexpress LLC HY14417A10MG

Encompass_Preferred

(1R,2S)-VU0155041 is the cis regioisomer of VU0155041 and a research-grade small molecule characterized as a partial agonist of metabotropic glutamate receptor 4 (mGluR4). It is intended for in vitro laboratory use and has documented agonist activity with EC50 ≈ 2.35 μM; additional allosteric modulatory activity (~0.74 μM) has also been reported.

  • IUPAC name: (1R,2S)-2-[(3,5-dichlorophenyl)carbamoyl]cyclohexane-1-carboxylic acid.
  • Partial agonist activity at mGluR4 with EC50 ≈ 2.35 μM.
  • Reported allosteric modulatory activity at ~0.74 μM in cell assays.
  • Purity approximately 95.1% as supplied.
  • Molecular formula C14H15Cl2NO3; molecular weight 316.18 g·mol-1.
  • Supplied as a research reagent for in vitro use only; not for human use.

Catalog No. 50-002-59117


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.