Learn More
Medchemexpress LLC (1R,2S)-xeruborbactam disodium | 2170836-14-1 | 99.3% | C10H8BFNa2O5 | 10MG
SDP
Supplier:  Medchemexpress LLC HY136072A10MG
Encompass_Preferred
(1R,2S)-Xeruborbactam disodium is a boronic acid derivative used as a research reagent and potent broad-spectrum β-lactamase inhibitor. Supplied as a dry powder for in vitro and analytical applications, it has a reported molecular formula of C10H8BFNa2O5 and molecular weight of 283.96 g·mol-1. Typical batch purity by LCMS is ~99.3%, and the compound is stored sealed at -20°C, protected from moisture.
  • Potent broad-spectrum β-lactamase inhibitor for mechanistic and resistance studies.
  • High purity suitable for analytical and bioassay use.
  • Solid powder for convenient storage and formulation.
  • Stable when stored sealed at -20°C, protected from moisture.
  • Available in small research-scale quantities for assay development.
Catalog No. 50-002-58796