Learn More
Medchemexpress LLC (1R,3R)-Ferroptosis inducer-1 | 2375357-97-2 | 99.3% | C25H21ClN2O5 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY145595A10MM/1ML
Encompass_Preferred
(1R,3R)-Ferroptosis inducer-1 is an isomer of Ferroptosis inducer-1 (HY-145595). Also known as compound BX-3a, it functions as a ferroptosis inducer with antitumor potential. This product is for research use only.
  • Isomer of Ferroptosis inducer-1
  • Functions as a ferroptosis inducer
  • Possesses antitumor potential
  • Available in solution (10 mM in DMSO)
Catalog No. 50-003-56216