Learn More
Medchemexpress LLC (1R,3R)-ferroptosis inducer-1 | 2375357-97-2 | 99.3% | C25H21ClN2O5 | 10MG

Supplier: Medchemexpress LLC HY145595A10MG
(1R,3R)-Ferroptosis inducer-1 is the (1R,3R) stereoisomer of a small-molecule ferroptosis inducer reported to promote ferroptotic cell death and to have antitumor potential in preclinical studies. Intended for laboratory research use, it is supplied as a white to off-white solid and is provided in solid and solution formats for experimental workflows.
- Induces ferroptosis and shows reported antitumor activity.
- Purity: 99.34%.
- Molecular formula: C25H21ClN2O5; molecular weight: 464.90.
- Available as a solid and as DMSO solutions for convenience.
- Supplied in multiple sizes to support small-scale and larger studies.
- CAS number: 2375357-97-2 for unambiguous identification.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.