Learn More
Medchemexpress LLC (1R,3R)-Ferroptosis inducer-1 | 2375357-97-2 | 99.34% | C25H21ClN2O5 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY145595A
Encompass_Preferred
(1R,3R)-Ferroptosis inducer-1 is an isomer of Ferroptosis inducer-1 (HY-145595), also known as compound BX-3a. This product functions as a Ferroptosis inducer with antitumor potential and is intended for research use only.
  • Isomer of Ferroptosis inducer-1
  • Functions as a Ferroptosis inducer
  • Possesses antitumor potential
  • For research use only
Catalog No. 50-003-53262