Learn More
Medchemexpress LLC (1R,3R)-RSL3 | 1219810-15-7 | >98.0% | 440.88 | C23H21ClN2O5 | 25 MG

Supplier: Medchemexpress LLC HY100218C25MG
(1R,3R)-RSL3 is the stereoisomer of RSL3 supplied as a research-grade powder for studies of ferroptosis and GPX4-related pathways. It is intended for in vitro use and is typically prepared as concentrated DMSO stock solutions following manufacturer guidance.
- Purity ≥98% (HPLC).
- Molecular weight 440.88.
- Chemical formula C23H21ClN2O5.
- CAS number 1219810-15-7.
- Appearance: powder; stable frozen for long-term storage.
- Solubility: DMSO up to 100 mg/mL; stock solution guidance provided for 1, 5, and 10 mM.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.