Learn More
Medchemexpress LLC (1R,3R)-RSL3 | 1219810-15-7 | ≥98.0% | 440.88 g/mol | C23H21ClN2O5 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY100218C100MG
Encompass_Preferred
(1R,3R)-RSL3 is the (1R,3R) stereoisomer of RSL3, a small-molecule ferroptosis activator that inhibits glutathione peroxidase 4 (GPX4). Supplied as a powder for research use, it is used to study ferroptosis and related cell-death pathways and is soluble in DMSO under gentle warming.
Catalog No. 50-002-15852