Learn More
Medchemexpress LLC 1R,3R-RSL3 | 1219810-15-7 | 99.5% | 440.88 | C23H21ClN2O5 | 10 MG

Supplier: Medchemexpress LLC HY100218C10MG
1R,3R-RSL3 is the enantiomeric isomer of RSL3, a small-molecule research compound used to investigate ferroptosis by modulating glutathione peroxidase 4 (GPX4). It is supplied as a solid research reagent with specified storage conditions and is intended for laboratory use in biochemical and cell-based assays.
- High analytical purity suitable for research applications.
- Molecular weight 440.88 for accurate dosing and calculations.
- CAS-registered chemical identifier for unambiguous reference.
- Recommended cold storage to preserve stability.
- Available in small-quantity packages for analytical and biological studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.