Learn More
Medchemexpress LLC (1R,3R)-RSL3 | 1219810-15-7 | 99.5% | 440.88 g·mol⁻¹ | C23H21ClN2O5 | 1 ML

Supplier: Medchemexpress LLC HY100218C10MM 1ML
1R,3R-RSL3 is the (1R,3R) stereoisomer of RSL3, a small-molecule inhibitor of glutathione peroxidase 4 (GPX4) used in research to induce ferroptosis. It is supplied as a powder and as a ready-to-use 10 mM solution in DMSO, and is characterized by defined storage conditions to preserve stability.
- High purity (99.5%) suitable for research applications.
- Available as powder or 10 mM solution in DMSO for convenience.
- Molecular weight 440.88 g·mol⁻¹ and formula C23H21ClN2O5.
- Soluble in DMSO at high concentration for stock preparation.
- Stable under recommended storage to maintain integrity.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.