Learn More
Medchemexpress LLC (1R,3R)-Rsl3 | 1219810-15-7 | 99.5% | 440.88 g/mol | C23H21ClN2O5 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY100218C50MG
Encompass_Preferred
(1R,3R)-RSL3 is the (1R,3R) stereoisomer of RSL3 supplied as an off-white to light yellow powder for research use. It modulates glutathione peroxidase 4 (GPX4) activity and is used to study ferroptotic cell death and related signaling pathways.
  • Off-white to light yellow solid for formulation and storage.
  • High purity (99.5%) suitable for research applications.
  • Stable when stored under recommended conditions for extended shelf life.
  • Available in multiple small pack sizes and ready-to-use DMSO solution formats.
  • Useful as a chiral control or comparator in ferroptosis assays.
Catalog No. 50-002-15323