Learn More
Medchemexpress LLC (1R,3R)-RSL3 | 1219810-15-7 | MFCD00038099 | ≥98.0% | 440.88 | C23H21ClN2O5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY100218C5MG
Encompass_Preferred
(1R,3R)-RSL3 is the 1R,3R stereoisomer of RSL3, a small-molecule research compound used in ferroptosis studies and related cell-based assays. Supplied as a dry powder in milligram quantities, it is intended for laboratory research use in biochemical and cellular experiments.
  • High purity suitable for research (≥98% by HPLC).
  • Powder form for flexible formulation and storage.
  • Stable under low-temperature storage (powder: -20°C; short-term at 4°C).
  • Well-characterized identifiers including CAS 1219810-15-7 and molecular weight 440.88.
  • Suitable for ferroptosis and apoptosis pathway studies in vitro.
Catalog No. 50-002-15297