Learn More
Medchemexpress LLC (1R,3S)-Compound E | 504428-17-5 | 99.6% | C27H24F2N4O3 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY14176A
Encompass_Preferred
(1R,3S)-Compound E is an isomer of Compound E (HY-14176) and serves as an experimental control. This compound functions as a γ-secretase inhibitor, effectively blocking the cleavage of β-amyloid (40), β-amyloid (42), and Notch γ-secretase, with IC50 values of 0.24, 0.37, and 0.32 nM respectively.
  • Isomer of compound E
  • Functions as an experimental control
  • Acts as a gamma-secretase inhibitor
  • Blocks beta-amyloid (40) cleavage
  • Blocks beta-amyloid (42) cleavage
  • Blocks notch gamma-secretase cleavage
  • IC50 values for cleavage are 0.24, 0.37, and 0.32 nM
Catalog No. 50-003-57077