Learn More
Medchemexpress LLC (1R,3S)-RSL3 | 1219810-13-5 | 99.8% | 440.88 | C23H21ClN2O5 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY100218B25MG
Encompass_Preferred
(1R,3S)-RSL3 is the less-active enantiomer of RSL3 used as a research reagent to study ferroptosis and GPX4-related mechanisms. In cellular assays it induces cell death in HT22 wild-type cells with an EC50 of 5.2 μM and is commonly used as a comparator to the more active enantiomer.
  • Chiral enantiomer used as a control in ferroptosis studies.
  • Induces cell death in HT22 cells (EC50 5.2 μM).
  • Molecular formula C23H21ClN2O5; molecular weight 440.88.
  • High purity (99.78%).
  • Supplied in small-mass quantities suitable for cellular assays.
Catalog No. 50-002-15628