Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (1R,3S)-RSL3 | 1219810-13-5 | 99.8% | 440.88 g/mol | C23H21ClN2O5 | 10 MG
SDP

Supplier:  Medchemexpress LLC HY100218B10MG

Encompass_Preferred

(1R,3S)-RSL3 is the less-active enantiomer of RSL3, supplied as a high-purity research reagent for studies of ferroptosis and GPX4-related pathways. It is provided as a solid for in vitro laboratory use with recommended cold storage to preserve stability.

  • Enantiomer of RSL3 used in ferroptosis research.
  • High purity: 99.8% by HPLC.
  • Molecular formula C23H21ClN2O5; molecular weight 440.88 g/mol.
  • CAS number 1219810-13-5 for unambiguous identification.
  • Supplied as a powder with cold storage recommendations for long-term stability.
  • Intended for laboratory research use only; not for human or clinical use.

Catalog No. 50-002-15825


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.