Learn More
Medchemexpress LLC (1R,3S)-RSL3 | 1219810-13-5 | 99.8% | 440.88 g/mol | C23H21ClN2O5 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY100218B100MG
Encompass_Preferred
(1R,3S)-RSL3 is the less active enantiomer of the ferroptosis activator RSL3 and is supplied for research use. It is a small-molecule modulator that interacts with glutathione peroxidase 4 (GPX4). Analytical documentation including datasheet, certificate of analysis, and safety data sheet is available from the manufacturer. Molecular formula C23H21ClN2O5; molecular weight 440.88 g/mol; CAS 1219810-13-5.
  • Less active enantiomer useful as a stereochemical control in experiments.
  • Modulates ferroptosis via interaction with GPX4.
  • High listed purity suitable for biochemical and cell-based assays.
  • Available in multiple small-scale pack sizes for research workflows.
  • Includes downloadable analytical documentation for data validation.
Catalog No. 50-002-15551