Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (1R,3S)-RSL3 | 1219810-13-5 | 99.8% | 440.88 g/mol | C23H21ClN2O5 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY100218B5MG

Encompass_Preferred

(1R,3S)-RSL3 is the less active enantiomer of RSL3, used in cellular studies of ferroptosis and GPX4-related pathways. It induces cell death in HT22 wild-type cells and is used as a stereochemical control to evaluate enantiomer-specific activity.

  • Less active enantiomer used as stereochemical control in ferroptosis studies
  • Reported cellular activity: EC50 5.2 μM in HT22 wild-type cells
  • High purity (≈99.78%) suitable for research applications
  • Solid powder with molecular formula C23H21ClN2O5 and molecular weight 440.88 g/mol
  • Stable under recommended storage: powder -20°C for long-term storage
  • Supplied as a small research quantity (5 mg) for laboratory use

Catalog No. 50-002-15406


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.