Learn More
Medchemexpress LLC (1R,3S)-RSL3 | 1219810-13-5 | 99.8% | 440.88 g·mol⁻¹ | C23H21ClN2O5 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY100218B50MG
Encompass_Preferred
(1R,3S)-RSL3 is the less active enantiomer of RSL3 provided for laboratory research. It induces cell death in HT22 wild-type cells in vitro (EC50 ≈ 5.2 μM) and is used as a tool compound to study ferroptosis and GPX4-related pathways.
  • Less active enantiomer of RSL3 used for comparative studies.
  • Research tool for studying ferroptosis and GPX4 inhibition.
  • High purity suitable for analytical and biological assays.
  • Available in small research-scale pack sizes for dosing flexibility.
  • Supplied as a powder with recommended cold storage for stability.
  • For research use only; not for human or clinical applications.
Catalog No. 50-002-15405