Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (1R)-tenofovir amibufenamide | 1571076-15-7 | 98.1% | 490.49 g·mol⁻¹ | C22H31N6O5P | 5 MG
SDP

Supplier:  Medchemexpress LLC HY137453B5MG

Encompass_Preferred

(1R)-Tenofovir amibufenamide is a tenofovir-class antiviral prodrug for preclinical and in vitro research. The compound is identified by CAS 1571076-15-7, has molecular formula C22H31N6O5P, and is used in studies of HBV and HIV. Intended for laboratory research use only; not for human or clinical use.

  • Antiviral tenofovir prodrug used in preclinical HBV and HIV research.
  • High purity (98.1%) suitable for analytical and biological assays.
  • Small research quantity supplied as 5 MG vial.
  • Molecular weight 490.49 g·mol⁻¹ and formula C22H31N6O5P.
  • Storage: pure form -20°C for long-term; in solvent -80°C for long-term.
  • Certificate of analysis and safety data available for verification.

Catalog No. 50-002-22944


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.