Learn More
Medchemexpress LLC (1S,2R)-1-Aminoindan-2-ol | 126456-43-7 | 149.19 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00253010G
Encompass_Preferred
(1S,2R)-1-Aminoindan-2-ol is a chiral resolving agent that efficiently resolves various racemic 2-arylalkanoic acids and precisely controls the stereostructure of products in asymmetric syntheses. It can also be used in the synthesis of HIV protease inhibitors.
  • Efficient chiral resolving agent
  • Resolves various racemic 2-arylalkanoic acids
  • Controls stereostructure in asymmetric syntheses
  • Used in synthesis of HIV protease inhibitors
Catalog No. 50-004-49503