Learn More
Strem, An Ascensus Company CAS# 135616-36-3. 1g. (1S,2S)-(+)-1,2-Cyclohexanediamino-N,N'-bis(3,5-di-t-butylsalicylidene), 98% (S,S)-Jacobsen Ligand. MFCD00191801
SDP
Supplier:  Strem, An Ascensus Company 0703171G
Encompass
CAS# 135616-36-3. 1g. (1S,2S)-(+)-1,2-Cyclohexanediamino-N,N'-bis(3,5-di-t-butylsalicylidene), 98% (S,S)-Jacobsen Ligand. MFCD00191801. Molecular Weight: 546.84. Molecular Formula: C36H54N2O2. Color/form: yellow pwdr. Strem# 07-0317. http://www.strem.com/catalog/v/07-0317/
Catalog No. 50-901-08958