Learn More
Strem, An Ascensus Company CAS# 188264-84-8. 1g. (1S,2S)-(+)-1,2-Cyclohexanediamino-N,N'-bis(3,5-di-t-butylsalicylidene)cobalt(II). MFCD01631278
SDP
Supplier:  Strem, An Ascensus Company 2705261G
Encompass
CAS# 188264-84-8. 1g. (1S,2S)-(+)-1,2-Cyclohexanediamino-N,N'-bis(3,5-di-t-butylsalicylidene)cobalt(II). MFCD01631278. Molecular Weight: 603.76. Molecular Formula: C36H52CoN2O2. Color/form: red-brown pwdr. Strem# 27-0526. http://www.strem.com/catalog/v/27-0526/
Catalog No. 50-901-11185