Learn More
Strem, An Ascensus Company CAS# 169689-05-8. 100mg. (1S,2S)-(-)-1,2-Diaminocyclohexane-N,N'-bis(2'-diphenylphosphinobenzoyl), 95% (S,S)-DACH-Phenyl Trost Ligand. MFCD01631274
SDP
Supplier:  Strem, An Ascensus Company 150961100MG
Encompass
CAS# 169689-05-8. 100mg. (1S,2S)-(-)-1,2-Diaminocyclohexane-N,N'-bis(2'-diphenylphosphinobenzoyl), 95% (S,S)-DACH-Phenyl Trost Ligand. MFCD01631274. Molecular Weight: 690.76. Molecular Formula: C44H40N2O2P2. Color/form: white to off-white pwdr. Strem# 15-0961. http://www.strem.com/catalog/v/15-0961/
Catalog No. 50-901-09989