Learn More
Medchemexpress LLC (1S,2S)-2-PCCA hydrochloride | 1609563-70-3 | 528.56 g/mol | C30H39Cl2N3O | 50 MG
SDP
Supplier:  Medchemexpress LLC HY100013B150MG
Encompass_Preferred
(1S,2S)-2-PCCA hydrochloride is the hydrochloride salt of the (1S,2S) diastereomer of 2-PCCA, a small-molecule ligand used to probe GPR88 receptor biology. It is intended for research use in biochemical and cell-based assays. Physical data: C30H39Cl2N3O; MW 528.56; CAS 1609563-70-3.
  • Hydrochloride salt form for improved solubility in DMSO.
  • Tool compound for investigating GPR88 receptor signaling in vitro.
  • Reported activity: related compound inhibits GPR88-mediated cAMP production (EC50 116 nM in HEK293 cells).
  • High solubility in DMSO (100 mg/mL; may require ultrasonic).
  • Suitable for biochemical, cellular, and analytical applications.
  • Recommended storage to preserve stability: 4°C for solid; in solution, -80°C up to 6 months.
Catalog No. 50-002-14995