Learn More
Medchemexpress LLC (1S,2S)-2-PCCA hydrochloride | 1609563-70-3 | 99.7% | 528.56 g/mol | C30H39Cl2N3O | 100 MG

Supplier: Medchemexpress LLC HY100013B1100MG
(1S,2S)-2-PCCA hydrochloride is the hydrochloride salt of the (1S,2S) diastereomer of 2-PCCA, a ligand active at the GPR88 GPCR. The (1S,2S) form is reported as less active than the (1R,2R) isomer; activity data for 2-PCCA indicate inhibition of GPR88-mediated cAMP production (EC50 ≈116 nM). This compound is provided for research use.
- High chemical purity (LCMS 99.65%).
- CAS number 1609563-70-3 for unambiguous identification.
- Molecular formula C30H39Cl2N3O; molecular weight 528.56 g/mol.
- Soluble in DMSO at 100 mg/mL (ultrasonic may be required).
- Storage recommendations: solid at 4°C under nitrogen; in solvent, -80°C (≤6 months) or -20°C (≤1 month).
- Intended for research use only; not validated for clinical use.
- Diastereomer is less potent than the corresponding (1R,2R) isomer (~5-fold difference reported).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.