Learn More
Medchemexpress LLC (1S,2S)-2-PCCA hydrochloride | 1609563-70-3 | 99.7% | 528.56 g/mol | C30H39Cl2N3O | 25 MG

Supplier: Medchemexpress LLC HY100013B125MG
(1S,2S)-2-PCCA hydrochloride is the less-active diastereomer of 2-PCCA, a small-molecule agonist of the GPR88 receptor. It inhibits GPR88-mediated cAMP production (EC50 = 116 nM in HEK293 cells) and is supplied as the hydrochloride salt for in vitro research use only.
- Hydrochloride salt form for increased stability.
- Reported GPR88 activity: EC50 116 nM (HEK293 cells).
- High purity (99.7%).
- Available in multiple small pack sizes for research workflows.
- Storage recommendations: 4°C; in solvent -80°C for 6 months, -20°C for 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.