Learn More
Medchemexpress LLC (1S,2S)-2-PCCA hydrochloride | 1609563-70-3 | 99.7% | 528.56 g/mol | C30H39Cl2N3O | 5 MG

Supplier: Medchemexpress LLC HY100013B15MG
(1S,2S)-2-PCCA hydrochloride is the (1S,2S) diastereomer of 2-PCCA supplied as the hydrochloride salt for research use. It is reported as a less active isomer compared with the active enantiomer and is used in studies of GPR88 receptor pharmacology and cAMP signaling in cell-based assays.
- Hydrochloride salt, suitable for biochemical and cell-based assays.
- CAS number 1609563-70-3.
- Molecular formula C30H39Cl2N3O; molecular weight 528.56 g/mol.
- Purity 99.7% (reported).
- Available as a solid or 10 mM solution in DMSO.
- Typical pack sizes include 5 mg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.